BAF188900100
Scienceware® rack slanter
polypropylene, pack of 1 ea
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.35
Produits recommandés
Matériaux
polypropylene
Caractéristiques
autoclavable
Holds 1 x rack
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18890-0100
Taille
293x242x83 mm
Catégories apparentées
Description générale
Tilt your test tube rack to 25°, 35° or 45°. Adjusting the angle of racks reduces hand strain when working with pipettors. Increase the surface area of your Agar inside test tubes. Fits racks up 293 mm x 242 mm x 83 mm H (11 9/16 x 9 9/16 x 3 1/4 in. H).
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP