Principaux documents
BAF187510160
Scienceware® POXYGRID® test-tube rack
Holds 60 x 10-13 mm tubes, blue
Synonyme(s) :
tube holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
blue
epoxy (coated steel)
Caractéristiques
Holds 60 x 10-13 mm tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18751-0160
L × l × H
17.5 cm × 11.4 cm × 6.4 cm
Description générale
Welded steel wire is coated with heat-cured, thermosetting epoxy to protect from rust and corrosion. The coating is unaffected by most organic solvents, weak acids and alkalis, and has excellent abrasion resistance and adhesion to steel substrate.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Informations légales
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP