Key Documents
930350
Sigma-Aldrich
TEM Grids
Carbon Film Supported Copper Hexagonal Mesh Grid, grid size 200 mesh, box of 50
Synonyme(s) :
TEM grids
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
product name
Carbon Film Supported Copper Hexagonal Mesh Grid, grid size 200 mesh, box of 50
Matériaux
carbon film
copper grid
Description
film layer thickness 5 - 6 nm carbon
Conditionnement
box of 50
Taille de la grille
200 mesh
Description générale
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.