Merck
"One-pot" fabrication of clickable monoliths for enzyme reactors.
Chemical communications (Cambridge, England) (2013-01-17)
Minghuo Wu, Hongquan Zhang, Zhixin Wang, Shengwen Shen, X Chris Le, Xing-Fang Li
RÉSUMÉ
A simple, novel "one-pot" approach was developed for the preparation of organic-silica hybrid monoliths. Two clickable monoliths functionalized with either azido or alkynyl moieties were prepared. The clickable monoliths were further used for the fabrication of enzyme reactors via "click chemistry".
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Pepsine from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsine from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsine from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsine from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsine from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsine from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Agarose−pepsine from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid