Key Documents
Z719404
Sorenson low binding aerosol barrier tips
MultiGuard E, maximum volume 10 μL
Synonyme(s) :
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
sterile
Caractéristiques
DNase free
RNase free
barrier
Conditionnement
pack of 96 ea (Racks)
Fabricant/nom de marque
Sorenson 15020T
Volume maximum
10 μL
Type de liaison
low binding surface
Description générale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.