Principaux documents
Z719463
Sorenson low binding aerosol barrier tips
MultiGuard, maximum volume 1000 μL
Synonyme(s) :
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Volume maximum:
1000 μL
Stérilité:
sterile
Caractéristiques:
barrier
Conditionnement:
pack of 1000 ea (10 racks of 100 tips)
Fabricant/nom de marque:
Sorenson 14200T
Matériaux
colorless
polypropylene
Stérilité
sterile
Caractéristiques
barrier
Conditionnement
pack of 1000 ea (10 racks of 100 tips)
Fabricant/nom de marque
Sorenson 14200T
Volume maximum
1000 μL
Description générale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.