Key Documents
EP5804719000
Eppendorf® Swing bucket rotor A-4-44
incl. conical form inserts
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 4 x buckets for 2 x 50 mL conical tubes
incl. conical form inserts
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5804719000
Paramètres
5,000 max. RCF (4,500 xg)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5804/5804R/5810/5810R
Informations légales
Eppendorf is a registered trademark of Eppendorf AG