Principaux documents
Z304204
Quickfit® column packing supports
fits ST/NS: 24/29 female joint
Synonyme(s) :
EX13/23
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Quickfit® EX 13/23
Disponibilité
not available in ROW (Available only in EU countries), USA and Canada
not available in USA and Canada
Diamètre
21 mm
Description générale
These spiral packing supports will hold 4 mm or larger Fenske helices. They may be used with larger socket sizes than those shown below providing there is a suitable constriction at or above the cone at the bottom of the column.
Informations légales
Quickfit is a registered trademark of SciLabware Limited