Principaux documents
Z188808
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 8-425, pkg of 1000 ea
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
black phenolic cap
white rubber liner
Caractéristiques
closure type black phenolic solid top cap
Conditionnement
pkg of 1000 ea
Diam. du tube de RMN
5 mm
Raccords
thread for 8-425
Description générale
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
These autoclavable caps are designed to ensure the ultimate in product safety.