Key Documents
Z174548
Sigma-Aldrich
Cow receiver for Aldrich® micro short-path distillation apparatus
joint: threaded 14/20
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
joint: threaded14/20
Catégories apparentées
Description générale
Removable fingers can hold approximately 12mL.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC