Key Documents
Z143952
Solid-glass beads
borosilicate, diam. 6 mm
Synonyme(s) :
borosilicate beads
About This Item
Code UNSPSC :
41102422
Nomenclature NACRES :
NB.43
Matériaux
borosilicate
Conditionnement
pack of 1 ea
pack of 4 ea
Diamètre
6 mm
Catégories apparentées
Description générale
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.