Principaux documents
Z106518
Supelco
Septa for open top caps
for cap size 15 mm, PTFE/silicone
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
Matériaux
PTFE/silicone
Conditionnement
pkg of 100
Description générale
For use with cut-out top caps and Wheaton vials.