Principaux documents
SYNA793524
Synthware bushing adapter
female joint: ST/NS 14/35, male joint: ST/NS 24/40
Synonyme(s) :
Synthware A793524, glass bushing adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
female joint: ST/NS14/35
male joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A793524
Catégories apparentées
Description générale
Bushing adapters allow for compact assemblies of dissimilar joints.
Informations légales
Synthware is a trademark of Kemtech America Inc.