Principaux documents
SYNA792445
Synthware bushing adapter
female joint: ST/NS 24/40, male joint: ST/NS 45/50
Synonyme(s) :
Synthware A792445, glass bushing adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
female joint: ST/NS24/40
male joint: ST/NS45/50
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A792445
Description générale
Bushing adapters allow for compact assemblies of dissimilar joints.
Informations légales
Synthware is a trademark of Kemtech America Inc.