Key Documents
SYNA551922
Synthware distillation distribution adapter
joint: ST/NS 19/22
Synonyme(s) :
Synthware A551922, cow distillation adapter
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A551922
Catégories apparentées
Description générale
Rotating cow receiver allows four flasks to be progressively filled.
Autres remarques
includes only the top part
Informations légales
Synthware is a trademark of Kemtech America Inc.