Key Documents
SYNA351420
Synthware distillation adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 14/20, side joint: 14/20, side arm angle: 75 deg
Synonyme(s) :
Synthware A351420, adapter, sidearm adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Joint
bottom joint: ST/NS14/20
side joint: 14/20
joint: ST/NS top14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A351420
Description générale
For use as a component in atmospheric or vacuum distillations.
Informations légales
Synthware is a trademark of Kemtech America Inc.