Principaux documents
SYNA341420
Synthware twin hose connecting adapter
joint: ST/NS 14/20
Synonyme(s) :
SynthwareA341420, Adapter, twin connecting hose
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A341420
D.E. du tuyau
8 mm
Catégories apparentées
Description générale
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Informations légales
Synthware is a trademark of Kemtech America Inc.