Principaux documents
BR703440
BRAND® Stand for Multichannel Transferpette® and Transferpette® S
Synonyme(s) :
Accessories, Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel Pipettes, Pipettes, Piston-operated pipettes, Single channel Pipettes
About This Item
Code UNSPSC :
41122801
Nomenclature NACRES :
NB.01
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 703440
Description générale
Individual stand for Transferpette®/Transferpette® S 8 channel / 12 channel. Holds 1 Transferpette® each.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This video explains how to adjust the volume on the Transferpette before being ready to pipette.