BR704807
BRAND® Benchtop Rack for 6 x Transferpette S
About This Item
Code UNSPSC :
41121508
Nomenclature NACRES :
NB.01
Produits recommandés
Caractéristiques
to hold 6 pipettes
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
BRAND 704807
Description générale
Save space in the laboratory and use stands to safely store your BRAND single or multichannel pipettes. BRAND has the right solution for every model and every need:- The single stand for the Transferpette® S mechanical pipette is usually used for the multichannel pipette.- Single stands are also available for the Transferpette® electronic pipette - one stand for pipettes up to 1000 µl and another for the 5 mL pipette.- The benchtop rack (carousel) stand holds up to six Transferpette® S or Transferpette® S -8/-12 pipettes, saving considerable space.- Three Transferpette® electronic pipettes (single channel, up to 1000 μl) can be charged simultaneously in the charging stand. Individual power supplies no longer needed!
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG