Skip to Content
Merck
All Photos(1)

Key Documents

C3222

Millipore

Campylobacter supplement III (Skirrow)

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

shipped in

wet ice

storage temp.

2-8°C

suitability

Campylobacter spp.

Application

An antibiotic supplement recommended for the selective isolation of Campylobacter species.

Components

Composition: (per vial)
Polymyxin B: 1,250 IU
Vancomycin: 5.00 mg
Trimethoprim: 2.50 mg

Reconstitution

Sufficient for 500 ml medium

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service