Skip to Content
Merck
All Photos(1)

Key Documents

17780

Millipore

Campylobacter Selective Supplement (Karmali)

suitable for microbiology

Synonym(s):

Karmali Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
veterinary

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

Campylobacter Selective Supplement (Karmali) is a selective supplement for isolating thermotolerant Campylobacter species from food and animal feeds.

Components

Composition (per vial; sufficient for 500 mL medium)
Sodium pyruvate 50 mg; Vancomycin 10 mg; Cefoperazon 16 mg; Cycloheximide 50 mg

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Okuno et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 286, 172-187 (2017-12-24)
Given its short hyperpolarization time (∼10

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service