Skip to Content
Merck
All Photos(2)

Key Documents

B2426

Millipore

Campylobacter Culture Medium

NutriSelect® Plus, according to ISO 10272-1:2017, powder, pH 7.4±0.2 (25 °C), bottle of 500 g, non-sterile, suitable for microbiology

Synonym(s):

Blood Free Campylobacter Selectivity Agar Base, Campylobacter Blood-Free Medium, Modified CCDA - Preston, Modified Charcoal Cefoperazone Desoxycholate Agar, mCCD agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

Agency

according to ISO 10272-1:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

bottle of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

selective for Campylobacter spp.

Looking for similar products? Visit Product Comparison Guide

Application

Used for selective isolation and differentiation of Campylobacter species.

Components

Components (g/L)
Beef extract 10.00
Peptic digest of animal tissue 10.00
Casein enzymic hydrolysate 3.00
Sodium chloride 5.00
Sodium deoxycholate 1.00
Ferrous sulphate 0.25
Sodium pyruvate 0.25
Charcoal, bacteriological 4.00
Agar 12.00

Preparation Note

Suspend 22.75 g of Blood Free Campylobacter Selectivity Agar Base in 500 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121 °C) for 15 minutes. Cool to 50°C and aseptically add 1 vial Blood Free Campylobacter Medium Selective Supplement (Cat. No. 74807) in 2.0 ml of distilled water, filtered through a sterile 0.4 μm filter to sterilize. Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

International Organization for Standardization, ISO/DIS 10272 (1995)

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service