Z227242
Union
PEEK, for connecting 1/16 in., bore size 0.25 mm
Synonyme(s) :
Unions Valco
About This Item
Produits recommandés
Matériaux
PEEK
Description
female
Taille de l'alésage
0.25 mm
Compatibilité
for connecting 1/16 in.
Choose from one of the most recent versions:
Certificats d'analyse (COA)