Documents
22999
Supelco
Unions Valco
internal reducing, bore 0.75 mm, for connecting 1/8 to 1/16 in.
Synonyme(s) :
Union réducteur interne
About This Item
Code UNSPSC :
41115700
Matériaux
stainless steel (nuts and ferrules included)
Conditionnement
pkg of 1 ea
Orifice
0.75 mm
Raccords
1/16 in. Valco
1/8 in. Valco
Compatibilité
for connecting 1/8 to 1/16 in.
Description générale
Acier inoxydable, sauf indication contraire. Écrous et ferrules inclus.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificats d'analyse (COA)