Principaux documents
Z741682
Sartorius low-retention pipette tips
volume range 0.5-200 μL, refill tower
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
Stérilité
non-sterile
Caractéristiques
DNase free
RNase free
barrier: no
endotoxin free
low retention
Conditionnement
pack of 960 ea (Refil)
Fabricant/nom de marque
Sartorius LH-L790202
Volume maximum
200 μL
Gamme de volumes
0.5-200 μL
Description générale
Sartorius Low Retention Tips have an extremely even and durable liquid-repellent surface for optimising the flow of the liquid while dispensing, for optimum sample recovery. Liquids with low surface tension, like detergents, slide easily off of the inner surface of the tip, leaving very little residue behind. This Low Retention feature is thus very useful in the following applications, where such viscous, detergent containing, liquids are often pipetted: PCR, real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods. The Ttps are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus, eLINE®, mLINE® and Proline® Plus pipettes. Tip trays are colour-coded for easy matching with corresponding, colour-coded Sartorius pipettes.
Autres remarques
Browse all pipette tips
Informations légales
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy