Principaux documents
Z740156
LifeSep magnetic separation stand
15SX, for 15 mL tubes
Synonyme(s) :
magnetic separation equipment
1 EA
874,00 $
874,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
874,00 $
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.56
874,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
for 15 mL tubes
Fabricant/nom de marque
Dexter Magnetic Technologies 2501023
Description générale
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Open-faced design for clear visibility
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
  • Durable and easy to use