Z710202
Nichipet Eco pipette
volume 1000-5000 μL
1 030,00 $
About This Item
1 030,00 $
Produits recommandés
Description
With glass tip
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Nichiryo 00-NPC-5000
Plage de mesure du volume
1000-5000 μL
Volume
5000 μL
Description générale
- Glass tip enables totally bias free liquid handling for endocrine disruptors analysis
- 4 sizes cover a wide volume range (20-10000 μL)
- Nozzle with double O-rings enables easy tip fitting/ejection
- Reliable sealing nozzle is PTFE and piston is ceramic