Principaux documents
Z659827
Bandelin SONOREX Ultrasonic bath accessories
tank lid for Z659800 and Z659045
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.74
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin D 514
Disponibilité
not available in USA and Canada
Description générale
When using chemically aggressive liquids (e.g. TICKOPUR J 80 U), removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath, use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Caractéristiques et avantages
  • for holding insert cups
  • sound-absorbing
  • shortens the heating phase
Informations légales
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG