BR759200
BRAND® UV cuvette micro
center H 8.5 mm, volume 70-850 μL, pack of 100 ea
Synonyme(s) :
Cuvettes, UV-Cuvettes
155,00 $
Disponible pour expédition le28 avril 2025Détails
About This Item
155,00 $
Disponible pour expédition le28 avril 2025Détails
Produits recommandés
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 759200
H centrale
8.5 mm
Volume
70-850 μL
Description générale
- BRAND® UV cuvette micro is manufactured under controlled room conditions.
- Ideal for the determinations in water analysis, chemistry, and life science applications.
- Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
- Clear, clean optical path with an indication of optical path orientation.
- Eliminates the risk of cross-contamination.
- Grouped by mold cavity number.
- An economical alternative to Quartz cuvettes.