Principaux documents
Z315001
Sigma-Aldrich
Hirschmann® pipetus® Junior pipette controller
for volumes 0.5 to 25 mL
Synonyme(s) :
Pipette controller
1 EA
286,00 $
286,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
286,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
286,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
plastic
silicone
Fabricant/nom de marque
Hirschmann 9902000
Volume
0.5-25 mL
Description générale
  • Low cost, hand-powered model
  • For volumes 0.5 to 25 mL
  • Fine dosing valve for accurate adjustment of meniscus
  • Discharge speed controlled by adjusting finger pressure on delivery trigger
  • Silicone pipette holder accepts volumetric, serological or Pasteur pipettes
  • Integral PTFE membrane filter prevents accidental ingress of liquid into controller
Informations légales
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Pipetus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Questions
  1. Is Hirschmann® pipetus® Junior pipette controller (catalog number Z315001) from Sigma-Aldrich rechargeable?
    1 answer
    1. No, this product is not rechargeable. This item is a manual pipette filler that does not have suction nor delivery that is motor driven. The suction and delivery are regulated by an internal bellows, which is inflated and deflated by the suction and delivery buttons. Please see the link to the rechargeable pipette controller below:
      https://www.sigmaaldrich.com/product/aldrich/z314951