Principaux documents
Z379255
Sigma-Aldrich
Freezebee ice bucket insert
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.22
Description générale
Fits standard sized ice bucket to keep samples cold and prevent their being lost in the bed of ice. Ingenious pattern permits use with 0.5 mL (48 each) or 1.5 mL (47 each) microcentrifuge tubes, 15 mL (8 each) or 50 mL (6 each) conical tubes.