Documents
Z353507
Sigma-Aldrich
Finnpipette® Multistepper® pipette tips
non-sterile, bag of bulk
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
turquoise
turquoise tip
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 400 ea (Packed in a bag)
bag of bulk
Fabricant/nom de marque
Finn 9401300
Volume maximum
1.5 mL
Gamme de volumes
1-1500 mL
Autres remarques
Browse all pipette tips
Informations légales
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Multistepper is a registered trademark of Labsystems