Key Documents
SMB00175
Sigma-Aldrich
Aromadendrin
≥95% (LC/MS-ELSD)
Synonyme(s) :
Dihydrokaempferol, Katuranin
About This Item
Formule empirique (notation de Hill):
C15H12O6
Numéro CAS:
Poids moléculaire :
288.25
Numéro MDL:
Code UNSPSC :
12352200
ID de substance PubChem :
Pureté
≥95% (LC/MS-ELSD)
Forme
solid
Application(s)
metabolomics
vitamins, nutraceuticals, and natural products
Température de stockage
−20°C
InChI
1S/C15H12O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,14-18,20H/t14-,15+/m0/s1
Clé InChI
PADQINQHPQKXNL-LSDHHAIUSA-N
Description générale
Natural product derived from plant source.
Actions biochimiques/physiologiques
Aromadendrin has been considered for potential use in the management of Type 2 diabetes due to its ability to stimulate glucose uptake and improve insulin resistance by inducing adipogenesis through increased PPAR2 expression.
Pictogrammes
Exclamation mark
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Acute Tox. 4 Oral
Code de la classe de stockage
11 - Combustible Solids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Les clients ont également consulté
Slide 1 of 2
1 of 2
Ken-ichi Morimura et al.
Phytochemistry, 67(24), 2681-2685 (2006-09-16)
From the leaves of Helicia cochinchinensis, collected on Okinawa Island, seven phenolic glucosides and two terpenic glucosides were isolated. Five of the phenolic glucosides were previously known, being identified with p-coumaric and ferulic acids glucosyl esters, rhodioloside, helicidiol, and naringenin
Izabela Redzynia et al.
Molecules (Basel, Switzerland), 14(10), 4147-4158 (2009-11-20)
X-ray structures of two compounds isolated from wood knots of coniferous trees, namely dihydrokaempferol (3,5,8,13-tetrahydroxyflavanon) and lariciresinol (3,14-dimetoxy-7,10-epoxylignan-4,15,19-triol), are presented here. Diffraction data for the Dihydrokaempferol crystals were collected on a CAD4 diffractometer and on a synchrotron for the lariciresinol
Wei Yun Zhang et al.
Pharmacology, 88(5-6), 266-274 (2011-11-08)
Agents that stimulate glucose uptake and improve insulin resistance may be useful in the management of type 2 diabetes mellitus (DM). Thus, the aims of this study were to assess the effects of aromadendrin, a flavonoid from Gleditsia sinensis Lam.
Wei Yun Zhang et al.
Biological & pharmaceutical bulletin, 33(9), 1494-1499 (2010-09-09)
The stimulation of glucose uptake into peripheral tissues is an important mechanism for the removal of glucose in blood and for the management of diabetes mellitus (DM). Since recent results have demonstrated the beneficial effects of flavonoids in relation to
Ilef Limem-Ben Amor et al.
Natural product communications, 5(5), 777-782 (2010-06-05)
Flavonoid hydroxylation is one way to increase the biological activities of these molecules and the number of hydroxyl groups needed for polymerization, esterification, alkylation, glycosylation and acylation reactions. These reactions have been suggested as a promising route to enhance flavonoid