PURXR
Pur-A-Lyzer™ Floating Racks for Dialysis
Maxi
Synonyme(s) :
Dialyzers, High through put dialysis tubing
Conditionnement
pack of 1 ea
Description générale
Floating rack can process up to 20 maxi Pur-A-Lyzer Dialysis Tubes in a single beaker.
Informations légales
Pur-A-Lyzer is a trademark of Sigma-Aldrich Co. LLC