Principaux documents
EP6001071110
Eppendorf® Side Access Stainless Steel Rack for CryoCube(R) F740 Upright Freezers
including front and back handle for storage of 48 Deepwell plates up to 86x128x53 mm, 3-Compartment, external W × D × H (139 mm) (569 mm) (444 mm)
1 EA
459,00 $
459,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
459,00 $
About This Item
Code UNSPSC :
41122800
459,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
stainless steel
Caractéristiques
3-Compartment
Holds 48 x deepwell plates
including front and back handle for storage of 48 Deepwell plates up to 86x128x53 mm
Fabricant/nom de marque
Eppendorf® 6001071110
Technique(s)
cryopreservation: suitable
Larg. externe × P × H
(139 mm) (569 mm) (444 mm)
Taille
549x139x444 mm
Description générale
Side access style stainless steel racks for Eppendorf CryoCube F740 ultra-low temperature upright freezers. By storing your sample inventory in an organized manner, you enable more economical use of the available space in your ULT freezer. Racks allow you to use 100% of the storage capacity. When using racks with side access, most of the storage boxes have to be pulled out of the cold interior of the ultra-low temperature freezer when accessing samples. In addition, metal racks have a long lifespan and can often be used for the next generation of ULT freezers. The stainless steel racks are assembled with 100% energy from renewable sources and made from 60-65% recycled steel. Coupled with Eppendorf′s use of cardboard packaging made from 90% recycled raw materials these are a good option to reduce your carbon footprint.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG