Principaux documents
EP5804754000
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer, Falcon and micro test tubes 7-17 mL, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
holds standard, Vacutainer, Falcon and micro test tubes 7-17 mL
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 5804754000
Technique(s)
centrifugation: suitable
Compatibilité
for use with 100 mL rectangular bucket
Description générale
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG