Principaux documents
EP0030128842
Eppendorf® Twin tec PCR plates, 96 wells
blue plate, conical bottom colorless wells, skirted, pkg of 300 ea
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Le produit EP0030128842 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
blue plate
conical bottom colorless wells
Caractéristiques
skirt
Conditionnement
pkg of 300 ea
Fabricant/nom de marque
Eppendorf® 0030128842
Technique(s)
PCR: suitable
Taille
96 wells
Description générale
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG