Principaux documents
EP0030128478
Eppendorf® Twin tec PCR plates, 96 wells
semi-skirted, with barcode, def. color (wells colorless), pkg of 25 ea
About This Item
Code UNSPSC :
41106300
Le produit EP0030128478 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Caractéristiques
skirt
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128478
Technique(s)
PCR: suitable
Taille
96 wells
Description générale
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG