Principaux documents
CLS6970SR
Corning® magnetic stir bar retrievers
ergonomic stir bar retriever, 1/cs
Synonyme(s) :
stir bar retrievers
1 EA
45,00 $
45,00 $
Disponible pour expédition le21 avril 2025Détails
Changer de vue
1 EA
45,00 $
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.73
45,00 $
Disponible pour expédition le21 avril 2025Détails
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
Corning 6970SR
Compatibilité
all stirring models
Catégories apparentées
Description générale
The Corning stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory. The container must be taken off the magnetic stirrer before using the retriever as the high-powered magnet in the stirrer cancels out the one in the retriever. The handle is designed for ergonomic and easy retrieval. The retriever is available one per case.
Informations légales
Corning is a registered trademark of Corning, Inc.