Documents
HS120553
Magnetic Stir Bar Retriever
white PTFE stir bar(s), L 300 mm
Synonyme(s) :
Magnetic Stirrer Retriever, Stir Bar Retriever
About This Item
Code UNSPSC :
41122401
Nomenclature NACRES :
NB.72
Matériaux
white PTFE stir bar(s)
Conditionnement
pack of 5 ea
Fabricant/nom de marque
Heathrow Scientific 120553
Longueur
300 mm
Description générale
The Stir Bar Retriever is encapsulated in PTFE making it easy to clean and durable.
Caractéristiques et avantages
  • The retrieval bar can recover stir bars even in hot solutions when stirring and mixing is no longer required
  • Chemically inert and easy to clean
  • Encapsulation in PTFE