Principaux documents
CLS3081200LO
Pyrex® large knob top desiccators
replacement desiccator bowl for 5.8L large knob top desiccator, 1/cs
Synonyme(s) :
desiccator bowls, desiccator lower bowls
1 EA
731,00 $
731,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
731,00 $
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
731,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
Corning 3081-200LO
Adéquation
suitable for (general analytical work)
Catégories apparentées
Description générale
Note: This is the replacement desiccator bowl for the Pyrex desiccator CLS3181200.
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
Informations légales
PYREX is a registered trademark of Corning, Inc.