Documents
Z803758
Specac® Omni Cell demountable cell for IR spectroscopy
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Fabricant/nom de marque
Specac Ltd
Technique(s)
IR spectroscopy: suitable
Description générale
Specac′s demountable Omni Cell System presents a novel approach for the analysis of liquid, mulls, pastes, and cast film samples in UV, Vis, NIR, and IR transmission spectroscopy, with one cell mount being suitable for all applications.
This transmission cell is specially designed to be suitable with both FTIR and dispersive instruments, and is available with a wide variety of high-quality window materials to meet your analytical needs.
Includes front and back plates, cell nest, 4 quick release nuts, bonded front PTFE gasket, rear neoprene gasket and 2 PTFE Luer plugs. Please order the preferred windows and spacers separately.
Informations légales
Specac is a registered trademark of Specac Ltd.
Pièce de rechange
Réf. du produit
Description
Tarif
Choose from one of the most recent versions:
Certificats d'analyse (COA)