Documents
Z800465
Specac® Omni Cell spacers
size 0.006 mm , rectangular
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Conditionnement
pkg of 5 ea
Fabricant/nom de marque
Specac Ltd
Technique(s)
IR spectroscopy: suitable
Taille
0.006 mm , rectangular
Informations légales
Specac is a registered trademark of Specac Ltd.
Choose from one of the most recent versions:
Certificats d'analyse (COA)