Principaux documents
UBBEL04UKC
Ubbelohde Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Size 1, Nom. Constant 0.01, Range 2 to 10 cSt
About This Item
Code UNSPSC :
41116107
Nomenclature NACRES :
NA.24
Fabricant/nom de marque
Paragon Scientific Ltd
Application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Description générale
This product is issued with a UKAS 17025 certificate of calibration