Documents
02712
SAFC
Cystamine dihydrochloride
Fabrication pharma
Synonyme(s) :
2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride
About This Item
Formule linéaire :
NH2CH2CH2SSCH2CH2NH2 · 2HCl
Numéro CAS:
Poids moléculaire :
225.20
Numéro Beilstein :
3616850
Numéro CE :
Numéro MDL:
Code UNSPSC :
12352100
Nomenclature NACRES :
NA.21
Source biologique
synthetic
Niveau de qualité
Pureté
≥98.0% (TLC)
Concentration
≥98 % (w/w) (T)
Impuretés
≤100 mg/kg 2,6-Di-tert-butyl-4-methylphenol (BHT) (HPLC)
≤100 ppm 1,2-dimethoxyethan
≤5000 ppm tert-butyl methyl ether
≤5000 ppm ethanol
Pf
215-225 °C (dec.)
217-220 °C (dec.) (lit.)
Solubilité
H2O: 1.0 g/10 mL, clear to almost clear, colorless to almost colorless (<5.5 NTU)
Adéquation
corresponds for H-NMR
Chaîne SMILES 
Cl[H].Cl[H].NCCSSCCN
InChI
1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H
Clé InChI
YUFRRMZSSPQMOS-UHFFFAOYSA-N
Pictogrammes
Exclamation mark
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Acute Tox. 4 Oral
Code de la classe de stockage
11 - Combustible Solids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Déjà en possession de ce produit ?
Yasue Ishii-Osai et al.
Journal of dermatological science, 67(1), 51-60 (2012-05-25)
N-propionyl-4-S-cysteaminylphenol (NPr-4-S-CAP) is selectively incorporated into melanoma cells and degrades them. However, it remains unclear whether NPr-4-S-CAP can induce cell death associated with the induction of host immune responses and tumor suppression in vivo. To examine the molecular mechanism of
Shosuke Ito et al.
Biochemical pharmacology, 84(5), 646-653 (2012-06-26)
Metastatic melanoma is resistant to conventional therapies. N-propionyl-4-S-cysteaminylphenol (NPrCAP), an N-protected sulfur-amine analog of tyrosine, is a good substrate for tyrosinase and is selectively incorporated into melanoma cells, causing cytotoxicity in vitro and in vivo. We have recently shown that
Thorsten Overath et al.
Molecular & cellular proteomics : MCP, 11(8), 467-477 (2012-05-05)
The post-translational modification of proteins with O-GlcNAc is involved in various cellular processes including signal transduction, transcription, translation, and nuclear transport. This transient protein modification enables cells or tissues to adapt to nutrient conditions or stress. O-Glycosylation of the 26
Zhongshan Liu et al.
Journal of chromatography. A, 1342, 70-77 (2014-04-15)
A polyhedral oligomeric silsesquioxane (POSS) hybrid monolith was simply prepared by using octaglycidyldimethylsilyl POSS (POSS-epoxy) and cystamine dihydrochloride as monomers via ring-opening polymerization. The effects of composition of prepolymerization solution and polycondensation temperature on the morphology and permeability of monolithic
Liang Gao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(11), 3185-3192 (2012-02-22)
Autofluorescent microcapsules were assembled by covalent cross-linking of polysaccharide alginate dialdehyde (ADA) derivative and cystamine dihydrochloride (CM) through a layer-by-layer (LBL) technique. The formulated Schiff base and disulfide bonds render capsules with pH- and redox-responsive properties for pinpointed intracellular delivery