Documents
Z303224
Quickfit® Liebig condenser
L 250 mm, joint: ST/NS 29/32
Synonyme(s) :
C1/14/25/SC
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/32
Fabricant/nom de marque
Quickfit C1/14/25/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Longueur
250 mm
Superficie
130 cm2
Description générale
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Informations légales
Quickfit is a registered trademark of SciLabware Limited