Key Documents
Z301302
Quickfit® plain bends
joint: ST/NS 14/23
Synonyme(s) :
RA9/11
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS14/23
Fabricant/nom de marque
Quickfit RA 9/11
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
Socket to cone. Generally used as distilling adapters.
Informations légales
Quickfit is a registered trademark of SciLabware Limited