Principaux documents
Z268518
Sigma-Aldrich
Vacushield
in-line filter designed to be placed in front of a vacuum pump, pkg of 3 ea
About This Item
Code UNSPSC :
41104900
Conditionnement
pkg of 3 ea
Fabricant/nom de marque
Gelman 4402
Description générale
Disposable, hydrophobic, in-line filter designed to be placed in front of a vacuum pump. Protects the pump from damage caused by aqueous solutions. Hose barb connector. Autoclavable.
Informations légales
Vacushield is a trademark of Pall Corporation