Principaux documents
Z268429
Sigma-Aldrich
Pall reusable syringe filter holders
diam. 25 mm
About This Item
Code UNSPSC :
41104900
eCl@ss :
32110206
Matériaux
polyacetal (resin construction)
polypropylene housing
Stérilité
non-sterile
Conditionnement
pkg of 6 ea
Fabricant/nom de marque
Gelman 4320
Diamètre
25 mm
Dimension de pores
(N/A)
Raccords
Luer inlet and outlet
Description générale
Reusable plastic filter holders provide an affordable alternative to disposable syringe filters. Autoclavable.