Principaux documents
Z256315
Tubing fittings combination set
more than 200 tubing clamps, hangers, wrap-it-tie cables
About This Item
Code UNSPSC :
41121815
Description générale
More than 200 tubing clamps, hangers, wrap-it-tie cables, and other organizers for laboratory tubing. Fittings are made of corrosion-resistant nylon or Delrin® plastic. Supplied in a PP box with hinged lid.
Informations légales
Delrin is a registered trademark of E. I. du Pont de Nemours and Company